Anti-FOXP1 Mouse Recombinant Antibody [Clone: rFOXP1/6902]
Catalog # 76967-574
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:12CC4
- Antigen Symbol:FOXP1
- Clonality:Recombinant
- Clone:rFOXP1/6902
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H334
- Antigen Synonyms:Forkhead box P1|FOXP1|FOX P1|FOXP 1|Fork head related protein like B|FLJ23741|Glutamine rich factor 1|Forkhead box protein P1|FOXP1_HUMAN
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 350-450, of human FOXP1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76967-574
- Packaging:Plastic vial
Specifications
About this item
Recombinant mouse monoclonal [rFOXP1/6902] antibody to FOXP1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: FOXP1
Clonality: Recombinant
Clone: rFOXP1/6902
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human