Anti-MCP2 Mouse Monoclonal Antibody [Clone: CCL8/3686]
Catalog # 76961-018
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:C C motif chemokine 8
- Antigen Symbol:MCP2
- Clonality:Monoclonal
- Clone:CCL8/3686
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P80075
- Antigen Synonyms:HC14|Chemokine (C C motif) ligand 8|MCP 2|MCP-2|CCL 8|Chemokine ligand 8|MCP-2(6-76)|Ccl8|CCL8_HUMAN
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:11 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 1-99, of human MCP2 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76961-018
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CCL8/3686] antibody to MCP2 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MCP2
Clonality: Monoclonal
Clone: CCL8/3686
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human