Anti-Angiotensin Converting Enzyme 1 Rabbit Recombinant Antibody [Clone: ACE/7004R]
Catalog # 76950-218
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ACE
- Antigen Symbol:Angiotensin Converting Enzyme 1
- Clonality:Recombinant
- Clone:ACE/7004R
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P12821
- Antigen Synonyms:ACE T|ACE1|Angiotensin I converting enzyme 1|Angiotensin Converting Enzyme 1|Angiotensin I converting enzyme|ACE_HUMAN|ACE 1|Angiotensin converting enzyme somatic isoform|Angiotensin converting enzyme testis specific isoform
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:150 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 652-788, of human Angiotensin Converting Enzyme 1.
- Purification:Protein A/G chromatography
- Cat. No.:76950-218
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [ACE/7004R] antibody to Angiotensin Converting Enzyme 1 for ELISA with samples derived from Human.
- Validated Applications: ELISA
Type: Primary
Antigen: Angiotensin Converting Enzyme 1
Clonality: Recombinant
Clone: ACE/7004R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human