Anti-PEA15 Mouse Monoclonal Antibody [Clone: 4D2]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal [4D2] antibody to PEA15 for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse and Bovine.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:1,000 to 1:2,000, ICC/IF: 1:1,000 to 1:2,000, IHC: 1:1,000 to 1:2,000
Type: Primary
Antigen: PEA15
Clonality: Monoclonal
Clone: 4D2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Bovine
Specifications
- Catalog No:
- 77073-890
- 77074-140
- Antigen Symbol:
- PEA15
- PEA15
- Antigen Name:
- 15 kDa phosphoprotein enriched in astrocytes
- 15 kDa phosphoprotein enriched in astrocytes
- Antigen Synonyms:
- HUMMAT 1H|Homolog of mouse MAT 1 oncogene|Astrocytic phosphoprotein PEA 15|Astrocytic phosphoprotein PEA15|Homolog of mouse MAT1 oncogene|HMAT 1|HMAT1|HUMMAT1H|Astrocytic phosphoprotein PEA-15
- HUMMAT 1H|Homolog of mouse MAT 1 oncogene|Astrocytic phosphoprotein PEA 15|Astrocytic phosphoprotein PEA15|Homolog of mouse MAT1 oncogene|HMAT 1|HMAT1|HUMMAT1H|Astrocytic phosphoprotein PEA-15
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 4D2
- 4D2
- Reactivity:
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# Q15121
- UniprotID# Q15121
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human PEA15, expressed in and purified from E. coli.
- Recombinant full-length human PEA15, expressed in and purified from E. coli.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 15 kDa
- 15 kDa
- Purification:
- Immunogen affinity purification
- Immunogen affinity purification
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate Buffered Saline with 50% Glycerol and 5mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested Applications:
- ICC/IF, IHC
- ICC/IF, IHC