To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Dual specificity mitogen activated protein kinase kinase 6
- Antigen Symbol:MEK6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P52564
- Antigen Synonyms:Dual specificity mitogen-activated protein kinase kinase 6|MAPKK6|MAP kinase kinase 6|MAP2K6|MEK6|MAPK/ERK kinase 6|MKK 6|MEK 6|MAPKK 6
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:37 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human MKK6 (amino acids 173-222)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77053-170
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to MKK6 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: MEK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat