Anti-AMPK alpha 1 + AMPK alpha 2 Rabbit Polyclonal Antibody
Catalog # 77052-296
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:5''-AMP-activated protein kinase catalytic subunit alpha-1
- Antigen Symbol:AMPK alpha 1 + AMPK alpha 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13131
- Antigen Synonyms:ACACA kinase|AMPK|AAPK1_HUMAN|AMPK alpha 1 + AMPK alpha 2|Acetyl-CoA carboxylase kinase|AMPK subunit alpha-1|AMPK alpha 2 chain|AMPK1|AMPK alpha 1 chain
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:62 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AMPK1 (amino acids 451-500)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77052-296
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to AMPK1 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: AMPK alpha 1 + AMPK alpha 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat