Anti-WASF2 Rabbit Polyclonal Antibody
77051-584
- Antibody Type:Primary
- Antigen Name:DICTYOSTELIUM
- Antigen Symbol:WASF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y6W5
- Antigen Synonyms:SCAR|Putative Wiskott Aldrich syndrome protein family member 4|SCAR2|Verprolin homology domain-containing protein 2|IMD2|Suppressor of cyclic-AMP receptor (WASP family)|SCAR 2|dJ393P12.2|Protein WAVE-2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:54 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human WASF2 (amino acids 141-190)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77051-584
- Packaging:Plastic vial
Rabbit polyclonal antibody to WASF2 for WB, IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: WASF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat