Anti-epithelial Sodium Channel gamma Rabbit Polyclonal Antibody
Catalog # 77051-392
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Amiloride sensitive epithelial sodium channel gamma subunit
- Antigen Symbol:epithelial Sodium Channel gamma
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P51170
- Antigen Synonyms:Epithelial Na(+) channel subunit gamma|Amiloride-sensitive sodium channel subunit gamma|ENaCG|ENaC gamma subunit|ENaCgamma|epithelial Sodium Channel gamma|BESC3|Epithelial Na+ channel subunit gamma|Amiloride sensitive sodium channel subunit gamma
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:74 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ENaC gamma (amino acids 132-181)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77051-392
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ENaC gamma for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: epithelial Sodium Channel gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat