Anti-BRCA1 Rabbit Polyclonal Antibody
Catalog # 77050-588
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:BRCA 1
- Antigen Symbol:BRCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P38398
- Antigen Synonyms:BRCA1/BRCA2 containing complex subunit 1|BRCA1/BRCA2-containing complex, subunit 1|BRCA1_HUMAN|BRCA1|BRCC 1|BRCAI|Breast and ovarian cancer susceptibility protein 1|BRCA1 DNA repair associated|BRCC1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:207 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human BRCA1 around the phosphorylation site of Ser1457 (amino acids 1423-1472)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77050-588
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to BRCA1 (phospho Ser1457) for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: BRCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human