Anti-TRF1 Rabbit Polyclonal Antibody
Catalog # 77050-532
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:hTRF1 AS
- Antigen Symbol:TRF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P54274
- Antigen Synonyms:t TRF1|NIMA interacting protein 2|PIN2|Telomeric protein Pin2/TRF1|PIN 2|Telomeric repeat binding factor 1|Telomeric repeat binding factor (NIMA interacting) 1|Telomeric protein Pin2|NIMA-interacting protein 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:50 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Telomeric Repeat Binding Factor 1 around the phosphorylation site of Ser219 (amino acids 185-234)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77050-532
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Telomeric Repeat Binding Factor 1 (phospho Ser219) for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: TRF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse