Anti-Cadherin 22 Rabbit Polyclonal Antibody
77049-536
- Antibody Type:Primary
- Antigen Name:C20orf25
- Antigen Symbol:Cadherin 22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9UJ99
- Antigen Synonyms:Cadherin-22|Cdh22|Ortholog of rat PB-cadherin|MGC39564|Cadherin-like 22|CAD22_HUMAN|dJ998H6.1|Cadherin 22, type 2|Cadherin 22
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:89 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human CDH22 (amino acids 111-160)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77049-536
- Packaging:Plastic vial
Rabbit polyclonal antibody to CDH22 for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: Cadherin 22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat