Anti-Brk Rabbit Polyclonal Antibody
77049-094
- Antibody Type:Primary
- Antigen Name:Breast tumor kinase
- Antigen Symbol:Brk
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13882
- Antigen Synonyms:Tyrosine protein kinase BRK|Tyrosine-protein kinase BRK|Breast tumour kinase|Protein-tyrosine kinase 6|Protein tyrosine kinase 6|PTK6_HUMAN|Brk|Ptk6
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:51 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Breast Tumor Kinase (amino acids 10-59)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77049-094
- Packaging:Plastic vial
Rabbit polyclonal antibody to Breast Tumor Kinase for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:20000
Type: Primary
Antigen: Brk
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse