Anti-KAT13C/NCOA2 Rabbit Polyclonal Antibody
Catalog # 77048-636
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:bHLHe75
- Antigen Symbol:KAT13C/NCOA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15596
- Antigen Synonyms:hTIF2|KAT13C/NCOA2|glucocorticoid receptor interacting protein 1|NCoA 2|MED1|GRIP 1|GRIP1|Class E basic helix-loop-helix protein 75|MGC138808
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:159 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NCoA2 around the phosphorylation site of Ser736 (amino acids 702-751)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77048-636
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NCoA2 (phospho Ser736) for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: KAT13C/NCOA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human