To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:E2AK4_HUMAN
- Antigen Symbol:GCN2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q9P2K8
- Antigen Synonyms:GCN2|Eukaryotic translation initiation factor 2-alpha kinase 4|GCN2-like protein|Eukaryotic Translation Initiation Factor 2 alpha kinase 4|GCN2 like protein|Eif2ak4|KIAA1338|MGCN2|GCN2 eIF2alpha kinase
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:186 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GCN2 around the phosphorylation site of Thr899 (amino acids 865-914)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77047-864
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GCN2 (phospho Thr899) for IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: GCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse