Anti-Histone H4R78me1 Rabbit Polyclonal Antibody
Catalog # 77047-712
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:H4F2
- Antigen Symbol:Histone H4R78me1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P62805
- Antigen Synonyms:Histone H4|HIST1H4|HISTH4H4|H4_HUMAN|HIST2H4|Hist4h4|Histone H4R78me1|H4FN|methyl histone H4
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:11 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Histone H4 around the acetylated site of Lys12 (amino acids 10-59)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77047-712
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Histone H4 (acetyl Lys12) for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:40000
Type: Primary
Antigen: Histone H4R78me1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat