Order Entry
United States
ContactUsLinkComponent
 
Anti-PRAK Rabbit Polyclonal Antibody
  77046-956
undefined
Anti-PRAK Rabbit Polyclonal Antibody
  77046-956
 :  A94603-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Host:
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Liquid
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Cat. No.:
    77046-956

 

 

Rabbit polyclonal antibody to MAPKAPK5 (phospho Thr182) for WB, IHC and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000

Type: Primary
Antigen: PRAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse