To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:epididymis luminal protein 55
- Antigen Symbol:Hsp20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# O14558
- Antigen Synonyms:Heat shock 20 kDa like protein p20|FLJ32389|Heat shock protein beta 6|Heat shock protein, 20-KD|HEL55|Heat shock protein alpha crystallin related B6|Heat-shock 27-KD protein 6|Heat shock 20 kDa-like protein p20|Heat shock protein beta-6
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:17 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human HSP20 around the phosphorylation site of Ser16 (amino acids 5-54)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77046-304
- Packaging:Plastic vial
Rabbit polyclonal antibody to HSP20 (phospho Ser16) for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: Hsp20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat