Anti-Notch2 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77042-682
Supplier: Antibodies.com
Supplier #: A93535-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to NOTCH2 (cleaved Val1697) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:100
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: Notch2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Clonality:Polyclonal
- Molecular Weight:83 kDa
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77042-682
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antibody Type:Primary
- Antigen Symbol:Notch2
- Gene ID:UniprotID# Q04721
- Immunogen:Synthetic peptide derived from human NOTCH2 (amino acids 1678-1727)
- Conjugation:Unconjugated
- Antigen Synonyms:Notch 2|N2ECD|Motch B|hN2|N2ICD|NOTC2_HUMAN|Notch Drosophila homolog 2|Notch 2 intracellular domain|neurogenic locus notch homolog protein 2
- Antigen Name:AGS2
- ELISA:Yes
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.



