Anti-beta III Tubulin Mouse Monoclonal Antibody [Clone: TU-20] (FITC (Fluorescein Isothiocyanate))
Catalog # 76951-158
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:beta 3 tubulin
- Antigen Symbol:beta III Tubulin
- Clonality:Monoclonal
- Clone:TU-20
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Hamster,Pig,Human,Bovine,Rat,Mouse,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13509
- Antigen Synonyms:M(beta)3|beta III Tubulin|CDCBM|beta-4|CFEOM3A|M(beta)6|MC1R|CDCBM1|CFEOM3
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Peptide (C) 441-448 coupled to maleimide-activated keyhole limpet hemocyanin via cysteine added to the N-terminus of the neuron-specific peptide
- Tested Applications:ICC
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76951-158
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TU-20] antibody to beta III Tubulin (FITC) for Flow Cytometry and ICC with samples derived from Mouse, Human, Rat, Hamster, Bovine, Feline and Porcine.
- Validated Applications: Flow Cytometry, ICC
- Recommended Dilutions: Flow Cytometry: 1-3 µg/ml
Type: Primary
Antigen: beta III Tubulin
Clonality: Monoclonal
Clone: TU-20
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Human, Rat, Hamster, Bovine, Cat, Porcine