To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:APO 3 ligand
- Antigen Symbol:TWEAK
- Clonality:Monoclonal
- Clone:CARL-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# O43508
- Antigen Synonyms:secreted form|MGC20669|MGC129581|APO3L|TNF-related weak inducer of apoptosis|APO3 ligand|APO 3L|APO3/DR3 ligand|DR3LG
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human CD255-transfected 2PK-3 cells
- Tested Applications:IHC
- Purification:Protein A chromatography
- Cat. No.:76964-276
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [CARL-1] antibody to TWEAK for Flow Cytometry and IHC with samples derived from Human.
- Validated Applications: Flow Cytometry, IHC
- Recommended Dilutions: Flow Cytometry: 1-4 µg/ml
Type: Primary
Antigen: TWEAK
Clonality: Monoclonal
Clone: CARL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 kappa
Reactivity: Human