Anti-CD26 Mouse Monoclonal Antibody [Clone: BA5b] (APC (Allophycocyanin))
76955-064
- Antibody Type:Primary
- Antigen Name:ADA-binding protein
- Antigen Symbol:CD26
- Clonality:Monoclonal
- Clone:BA5b
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# P27487
- Antigen Synonyms:ADCP 2|CD26 antigen 3|ADCP2|Adenosine deaminase complexing protein 2|CD 26|ADABP|CD26 antigen|ADCP-2|CD26
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:A human T cell clone
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76955-064
- Packaging:Glass bottle
Mouse monoclonal [BA5b] antibody to CD26 (APC) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD26
Clonality: Monoclonal
Clone: BA5b
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human