To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:269C wild type
- Antigen Symbol:Integrin alpha 4
- Clonality:Monoclonal
- Clone:9F10
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Horse,Sheep,Non-human primates,Human,Bovine,Dog,Cat
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P13612
- Antigen Synonyms:CD49d|Antigen CD49D, alpha 4 subunit of VLA 4 receptor|CD49 antigen-like family member D|CD49 antigen like family member D|Integrin alpha IV|IA4|Integrin alpha-4|Integrin alpha 4 subunit|Integrin alpha 4
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 15 mM Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Tested Applications:IHC-Fr
- Purification:Protein A chromatography
- Cat. No.:76965-660
- Packaging:Plastic vial
Mouse monoclonal [9F10] antibody to Integrin alpha 4 for Flow Cytometry and IHC-Fr with samples derived from Human, Bovine, Canine, Horse, Feline, Sheep and Non-Human Primates.
- Validated Applications: Flow Cytometry, IHC-Fr
- Recommended Dilutions: IHC: 5-10 µg/ml
Type: Primary
Antigen: Integrin alpha 4
Clonality: Monoclonal
Clone: 9F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Bovine, Dog, Horse, Cat, Sheep, Non-Human Primates