Anti-Integrin beta 3 Mouse Monoclonal Antibody [Clone: VIPL2] (FITC (Fluorescein Isothiocyanate))
76949-242
- Antibody Type:Primary
- Antigen Name:BDPLT16
- Antigen Symbol:Integrin beta 3
- Clonality:Monoclonal
- Clone:VIPL2
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Size:100 T
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen Synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76949-242
- Packaging:Glass bottle
Mouse monoclonal [VIPL2] antibody to Integrin beta 3 (FITC) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: VIPL2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates