About this item
Rabbit polyclonal antibody to GAPex 5 for WB and ICC/IF with samples derived from Mouse.
- Validated Applications: WB, ICC/IF
- Recommended Dilutions: WB: 1:500-1:2, 000, ICC/IF: 1:50-1:100
Type: Primary
Antigen: GAPex 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77202-174
- Antigen Symbol:
- GAPex 5
- Antigen Name:
- GAPex 5
- Antigen Synonyms:
- GTPase activating protein and VPS9 domains 1|Rab5 activating protein 6|KIAA1521|RAP6|GAPEX5|GAPVD1
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q14C86
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human GAPVD1 (NP_056450.2)
- Sequence:
- MVKLDIHTLAHHLKQERLYVNSEKQLIQRLNADVLKTAEKLYRTAWIAKQQRINLDRLIITSAEASPAECCQHAKILEDTQFVDGYKQLGFQETAYGEFLSRLRENPRLIASSLVAGEKLNQENTQSVIYTVFTSLYGNCIMQEDESYLLQVLRYLIEFELKESDNPRRLLRRGTCAFSILFKLFSEGLFSAKLFLTATLHEPIMQLLVEDEDHLETDPNKLIERFSPSQQEKLFGEKGSDRFRQKVQEMVESNEAKLVA
- Form:
- Liquid
- Molecular Weight:
- 165 kDa
- Purification:
- Affinity purification
- Storage Buffer:
- Supplied in Phosphate Buffered Saline, pH 7.3, with 50% Glycerol and 0.01% Thiomersal
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Tested Applications:
- ICC/IF