Specifications
- Pack type:Vial
- Description:Endotoxin specific buffer, beta glucan blocker, xact formulation previously distributed by Charles River Endosafe under Cat. No. BG120
- Size:6×5.3 ml
- Cat. No.:76994-334
- Application:Ideal for LAL that has the potential to react with Beta Glucans
Specifications
About this item
Our Endotoxin Test-Related Product are additional specialty kits that assist with the successful completion of bacterial endotoxin tests.
- Endotoxin extracting solution: Capable of extracting endotoxins that are unable to be extracted in water or saline, Recommended for use in endotoxin testing on equipment and devices that may come into contact with fluids containing blood or protein
- ES Buffer: Each vial has solution for use in reconstituting a maximum of 5.2 ml of reagent, Ideal for LAL that has the potential to react with glucans,thus making an endotoxinspecific reagent
- Control Standard Endotoxin: Endotoxin derived from E. coli UKT-B, Can be used to prepare controls and standard curves, Potency of CSE supplied to match each lot of LAL, Reconstituted CSE can be stored at 2 to 10 °C for one month
- LPS: E. coli O55:B5 (>100,000 EU/vial), Can make multiple endotoxin indicators from one vial, Used for validation studies
PS Single Test: Endotoxin-specific reagent avoids false positive results from glucans, This kit is in single-test configuration to avoid contamination, KTA quantitation for this product is performed on the Toxinometer® • Allows endotoxin-specific measurement in samples that typically have inhibitory components: ethanol, silicone oil, soybean oil, olive oil, stearic acid, egg yolk lecithin, oiladjuvanted vaccine, albumin products, coagulation factor products (factor 8), interferon formulation, antithrombin III, Immunoglobulin product and other ethanol-soluble samples.
Available to address special testing needs.