Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/520]
Catalog # 76953-258
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ADMCKD
- Antigen Symbol:MUC1
- Clonality:Monoclonal
- Clone:MUC1/520
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a lambda
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P15941
- Antigen Synonyms:CA 15-3|ADMCKD1|Carcinoma associated mucin|CA15.3|CA15 3|Breast carcinoma associated antigen DF3|CA15 3 antigen|Cancer antigen 15-3|Breast carcinoma-associated antigen DF3
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:265 - 400 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Human milk fat globule membranes
- Tested Applications:IHC
- Purification:Protein A/G chromatography
- Cat. No.:76953-258
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MUC1/520] antibody to MUC1 for IHC with samples derived from Human.
- Validated Applications: IHC
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: MUC1/520
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human