Anti-Integrin alpha E Rabbit Recombinant Antibody [clone: ITGAE/3904R]
76956-616
- Antibody Type:Primary
- Antigen Name:A530055J10
- Antigen Symbol:Integrin alpha E
- Clonality:Recombinant
- Clone:ITGAE/3904R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P38570
- Antigen Synonyms:alpha-M290|aM290|alpha-E1|HML 1 antigen|CD 103|Antigen CD103|HML-1 antigen|CD103 antigen|HML1 antigen
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:150 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment of human ITGAE protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76956-616
- Packaging:Plastic vial
Recombinant rabbit monoclonal [ITGAE/3904R] antibody to Integrin alpha E for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin alpha E
Clonality: Recombinant
Clone: ITGAE/3904R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human