To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:AACT
- Antigen Symbol:alpha 1 Antichymotrypsin
- Clonality:Monoclonal
- Clone:AACT/1452
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01011
- Antigen Synonyms:Growth inhibiting protein 24|Cell growth inhibiting gene 24/25 protein|alpha 1 Antichymotrypsin|Cell growth-inhibiting gene 24/25 protein|Alpha-1-antichymotrypsin His-Pro-less|Growth inhibiting protein 25|GIG24|GIG25|AACT_HUMAN
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:65 - 76 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 49-187, of human Antichymotrypsin protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76968-170
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [AACT/1452] antibody to alpha 1 Antichymotrypsin for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: alpha 1 Antichymotrypsin
Clonality: Monoclonal
Clone: AACT/1452
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human