To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cit Histone H3
- Antigen Symbol:Histone H3
- Clonality:Recombinant
- Clone:PHH3/471R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P68431
- Antigen Synonyms:H3 3 like sequence MH921|FLJ92264|H3 a|citrullin|H3 3A|citrulline|Citrullated|H 3|Citrullinated Histone H3
- Modification:phospho Ser10
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:15 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Synthetic peptide corresponding to amino acids within 1-100 of Histone H3 (phospho S10)
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76961-996
- Packaging:Plastic vial
Specifications
About this item
Recombinant rabbit monoclonal [PHH3/471R] antibody to Histone H3 (phospho Ser10) for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Histone H3
Clonality: Recombinant
Clone: PHH3/471R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human