To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:A I
- Antigen Symbol:Arginase 1
- Clonality:Monoclonal
- Clone:ARG1/1125 + ARG1/1126
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:Multiple
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P05089
- Antigen Synonyms:Al|ARG 1|Arginase-1|Arginase 1|Arginase type I|ARGI1_HUMAN|Arginase, liver|Arginase liver|arg1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:35 - 38 kDa
- Storage Temperature:Shipped at 4 °C. Store at 4 °C.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant fragment, around amino acids 11-97, of human ARG1 protein. The exact sequence is proprietary.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76968-882
- Packaging:Plastic vial
Mouse monoclonal [ARG1/1125 + ARG1/1126] antibody to Arginase 1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: Arginase 1
Clonality: Monoclonal
Clone: ARG1/1125 + ARG1/1126
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human