To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Annexin 1
- Antigen Symbol:Annexin A1
- Clonality:Monoclonal
- Clone:CPTC-ANXA1-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P04083
- Antigen Synonyms:ANX 1|Annexin I (lipocortin I)|Annexin I|ANX1|ANX A1|Annexin-1|AnnexinA1|AnnexinI|Annexin A1
- Storage Buffer:Supplied in 10 mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:38 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant full-length human Annexin A1 protein
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76959-864
- Packaging:Plastic vial
Mouse monoclonal [CPTC-ANXA1-1] antibody to Annexin A1 for IP, WB and IHC-P with samples derived from Human.
- Validated Applications: IP, WB, IHC-P
- Recommended Dilutions: IP: 2 to 4 µg / mg protein, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Annexin A1
Clonality: Monoclonal
Clone: CPTC-ANXA1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human