To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:CD172 antigen like family member B
- Antigen Symbol:CD172b
- Clonality:Monoclonal
- Clone:B4B6
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Size:100 tests
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# O00241
- Antigen Synonyms:SIRP BETA 1|CD172b antigen|1/CD172b|Signal regulatory protein beta 1|SIRPß|DKFZp686A05192|SIRP beta 1 isoform 3|CD172b
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:NIH-3T3 cells transfected with human CD172b
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76958-584
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [B4B6] antibody to CD172b (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD172b
Clonality: Monoclonal
Clone: B4B6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human