Anti-CD158a/g/h Mouse Monoclonal Antibody [clone: HP-MA4] (PE (Phycoerythrin))
Catalog # 76966-530
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD158a/g/h
- Clonality:Monoclonal
- Clone:HP-MA4
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Size:100 tests
- Form:Liquid
- Gene ID:UniprotID# Q99706
- Storage Buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage Temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Human NK cell line LB2
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. No.:76966-530
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [HP-MA4] antibody to CD158a/g/h (PE) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD158a/g/h
Clonality: Monoclonal
Clone: HP-MA4
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human