Anti-Collagenase ColA Mouse Monoclonal Antibody [clone: cp-02]
77080-568
- Antibody Type:Primary
- Antigen Symbol:Collagenase ColA
- Clonality:Monoclonal
- Clone:cp-02
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Clostridium perfringens,Clostridium histolyticum
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P43153,Q46085
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol (filter sterilised, without Sodium Azide, and carrier free).
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C.
- Immunogen:Culture supernatant of Clostridium perfringens
- Purification:Affinity purification
- Cat. No.:77080-568
- Packaging:Plastic vial
Mouse monoclonal (cp-02) antibody to Clostridium perfringens Collagenase ColA for WB and ELISA with samples derived from Clostridium perfringens and Clostridium histolyticum.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1,000
Type: Primary
Antigen: Collagenase ColA
Clonality: Monoclonal
Clone: cp-02
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Clostridium perfringens, Clostridium histolyticum