Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-Thymidine Kinase 1 Rabbit Polyclonal Antibody
Anti-Thymidine Kinase 1 Rabbit Polyclonal Antibody
Catalog # 77062-698
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Thymidine Kinase 1 Rabbit Polyclonal Antibody
Catalog # 77062-698
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A101406-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    cytosolic
  • Antigen Symbol:
    Thymidine Kinase 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04183
  • Antigen Synonyms:
    KITH_HUMAN|TK1|Thymidine kinase 1 soluble isoform|TK 1|Thymidine Kinase 1|Thymidine kinase cytosolic|Thymidine kinase|TK 2|Thymidine kinase 1 soluble
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    25 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at +4 °C.
  • Immunogen:
    Synthetic peptide derived from human KITH (amino acids 144-193)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. No.:
    77062-698
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to KITH for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: Thymidine Kinase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human