Anti-CLECSF6 Rabbit Polyclonal Antibody
100 µg, Plastic vial
77076-820
: A99306-100
Rabbit polyclonal antibody to CLEC4A for WB, IHC and ELISA with samples derived from Human.
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC to P: 1:50 to 1:300, ELISA: 1:10000
: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: CLECSF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Size:100 µg
- Packaging:Plastic vial
- Clonality:Polyclonal
- Molecular Weight:27 kDa
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol.
- Tested Applications:IHC
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77076-820
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antibody Type:Primary
- Antigen Symbol:CLECSF6
- Gene ID:UniprotID# Q9UMR7
- Immunogen:Synthetic peptide derived from the C-terminal region of human CLEC4A (amino acids 161-210)
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:C-type lectin superfamily member 6|C type lectin DDB27|C-type lectin DDB27|C type lectin|C type lectin superfamily 6|C-type lectin domain family 4, member A|C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6|C-type lectin domain family 4, member a2|C-type lectin domain family 4 member A
- Antigen Name:C type (calcium dependent carbohydrate recognition domain) lectin superfamily member 6
- ELISA:Yes
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



