Anti-Apolipoprotein A I Rabbit Polyclonal Antibody
77066-716
- Antibody Type:Primary
- Antigen Name:Apo-AI
- Antigen Symbol:Apolipoprotein A I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P02647
- Antigen Synonyms:Apolipoprotein A-I|ApoA-I|APOA1_HUMAN|Apolipoprotein A I|Apolipoprotein AI|Apolipoprotein A1|Apolipoprotein A-I(1-242)|ApoA I|APOA1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:30 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the Internal region of human APOA1 (amino acids 81-130)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77066-716
- Packaging:Plastic vial
Rabbit polyclonal antibody to APOA1 for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Apolipoprotein A I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse