To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:GLP
- Antigen Symbol:GLP1R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# P43220
- Antigen Synonyms:GLP-1-R|GLP 1R|GLP 1 receptor|Glucagon like peptide 1 receptor|GLP1R|GLP-1 receptor|GLP1R_HUMAN|GLP-1R|GLP 1 R
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:53 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GLP1R (amino acids 101-150)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77063-314
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to GLP1R for IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: GLP1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse