Anti-ATF3 [CL1685] Rabbit Polyclonal Antibody
Catalog # 77067-064
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:Activating transcription factor 3
- Antigen Symbol:ATF3 [CL1685]
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P18847
- Antigen Synonyms:cAMP dependent transcription factor ATF3|Cyclic AMP dependent transcription factor ATF3|cAMP-dependent transcription factor ATF-3|ATF3deltaZip2c|ATF3 [CL1685]|ATF3|ATF3_HUMAN|ATF3deltaZip2|ATF3deltaZip3
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:20 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ATF3 (amino acids 131-180)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77067-064
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ATF3 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: ATF3 [CL1685]
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat