Anti-EDG5 Rabbit Polyclonal Antibody
77061-300
- Antibody Type:Primary
- Antigen Name:AGR16
- Antigen Symbol:EDG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O95136
- Antigen Synonyms:Endothelial differentiation G-protein coupled receptor 5|H218|Endothelial differentiation sphingolipid G protein coupled receptor 5|Endothelial Cell Differentiation Gene 5|EDG 5|EDG5|Endothelial differentiation G protein coupled receptor 5|LPb2|Gpcr13
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:38 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human EDG5 (amino acids 261-310)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77061-300
- Packaging:Plastic vial
Rabbit polyclonal antibody to EDG5 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: EDG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat