Anti-RNA terminal phosphate cyclase-like 1 Rabbit Polyclonal Antibody
77060-334
- Antibody Type:Primary
- Antigen Name:RCL1
- Antigen Symbol:RNA terminal phosphate cyclase-like 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9Y2P8
- Antigen Synonyms:RNA 3' terminal phosphate cyclase like protein|RNA terminal phosphate cyclase-like 1|RNAC|RPC2|RNA terminal phosphate cyclase like 1|RNA 3''-terminal phosphate cyclase-like protein|RNA cyclase homolog|RPCL1|RCL1_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RCL1 (amino acids 307-356)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77060-334
- Packaging:Plastic vial
Rabbit polyclonal antibody to RCL1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: RNA terminal phosphate cyclase-like 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat