Anti-eIF4EBP1 Rabbit Polyclonal Antibody
Catalog # 77059-878
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:4E-BP1
- Antigen Symbol:eIF4EBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q60876
- Antigen Synonyms:PHASI|BP 1|4EBP1_HUMAN|PHAS-I|eIF4E binding protein 1|eIF4E-binding protein 1|4EBP1|eIF4EBP1|Eukaryotic translation initiation factor 4E-binding protein 1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:12 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from mouse 4E-BP1 (amino acids 35-84)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77059-878
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to 4E-BP1 for WB, IHC and ELISA with samples derived from Mouse and Rat.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:5000
Type: Primary
Antigen: eIF4EBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat