Anti-ACSS1 Rabbit Polyclonal Antibody
Catalog # 77059-534
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ACAS2L
- Antigen Symbol:ACSS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NUB1
- Antigen Synonyms:Acetyl CoA synthetase 2|Acetyl Coenzyme A synthetase 2 (AMP forming) like|AceCS2L|AceCS2|Acetyl coenzyme A synthetase 2 like, mitochondrial|Acetate--CoA ligase 2|Acetate CoA ligase 2|Acetyl-coenzyme A synthetase 2-like, mitochondrial|Acetyl-CoA synthetase 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:74 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ACSS1 (amino acids 611-660)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77059-534
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ACSS1 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: ACSS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse