Anti-Aldolase C Rabbit Polyclonal Antibody
Catalog # 77059-434
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ALDC
- Antigen Symbol:Aldolase C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P09972
- Antigen Synonyms:aldoc|Brain-type aldolase|Aldolase C|Brain type aldolase|ALDO C|ALDOC_HUMAN|Aldolase 3|Fructoaldolase C|Aldolase C Fructose bisphosphate
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:39 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from the N-terminal region of human ALDOC (amino acids 21-70)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Cat. No.:77059-434
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ALDOC for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Aldolase C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat