Anti-PEA15 Rabbit Polyclonal Antibody
Catalog # 77057-958
Supplier: Antibodies.com
Specifications
- Antibody Type:Primary
- Antigen Name:15 kDa phosphoprotein enriched in astrocytes
- Antigen Symbol:PEA15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q15121
- Antigen Synonyms:HUMMAT 1H|Homolog of mouse MAT 1 oncogene|Astrocytic phosphoprotein PEA 15|Astrocytic phosphoprotein PEA15|Homolog of mouse MAT1 oncogene|HMAT 1|HMAT1|HUMMAT1H|Astrocytic phosphoprotein PEA-15
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:15 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PEA15 (amino acids 61-110)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77057-958
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to PEA15 for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:1000
Type: Primary
Antigen: PEA15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat