Anti-1-AGP acyltransferase 4 Rabbit Polyclonal Antibody
Catalog # 77056-422
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:1 acyl sn glycerol 3 phosphate acyltransferase delta
- Antigen Symbol:1-AGP acyltransferase 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NRZ5
- Antigen Synonyms:1-acylglycerol-3-phosphate O-acyltransferase 4|1-AGP acyltransferase 4|1 AGP acyltransferase 4|1 acylglycerol 3 phosphate O acyltransferase 4|dJ473J16.2|1-AGPAT 4|1-acylglycerol-3-phosphate O-acyltransferase 4 (lysophosphatidic acid acyltransferase, delta)|1-acyl-sn-glycerol-3-phosphate acyltransferase delta|1 AGPAT4
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:44 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human AGPAT4 (amino acids 140-189)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77056-422
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to AGPAT4 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: 1-AGP acyltransferase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat