Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-26S proteasome non-ATPase regulatory subunit 12 Rabbit Polyclonal Antibody
Anti-26S proteasome non-ATPase regulatory subunit 12 Rabbit Polyclonal Antibody
Catalog # 77055-638
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-26S proteasome non-ATPase regulatory subunit 12 Rabbit Polyclonal Antibody
Catalog # 77055-638
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A96772-100

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    26S proteasome non-ATPase regulatory subunit 12
  • Antigen Symbol:
    26S proteasome non-ATPase regulatory subunit 12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O00232
  • Antigen Synonyms:
    MGC75406|proteasome (prosome, macropain) 26S subunit, non ATPase12|PSMD12|26S proteasome regulatory subunit p55|26S proteasome regulatory subunit RPN5|PSD12_HUMAN|Proteasome 26S non ATPase subunit 12 isoform 2
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    52 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human PSMD12 (amino acids 151-200)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Cat. No.:
    77055-638
  • Packaging:
    Plastic vial

Specifications

About this item

Rabbit polyclonal antibody to PSMD12 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000

Type: Primary
Antigen: 26S proteasome non-ATPase regulatory subunit 12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat