Anti-CREB3L2 Rabbit Polyclonal Antibody
100 µg, Plastic vial
Catalog # 77054-886
Supplier: Antibodies.com
Supplier #: A96584-100
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to CREB3L2 for WB and ELISA with samples derived from Human, Mouse and Rat.
Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Type: Primary
Antigen: CREB3L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Documents
Specifications
- Size:100 µg
- Packaging:Plastic vial
- Clonality:Polyclonal
- Molecular Weight:57 kDa
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Form:Liquid
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77054-886
- Reactivity:Human|Mouse|Rat
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice at 4 °C
- Antibody Type:Primary
- Antigen Symbol:CREB3L2
- Gene ID:UniprotID# Q70SY1
- Immunogen:Synthetic peptide derived from human CREB3L2 (amino acids 250-299)
- Conjugation:Unconjugated
- Antigen Synonyms:cAMP responsive element binding protein 3 like 2|BBF2H7|BZIB transcription factor|cAMP-responsive element-binding protein 3-like protein 2|Basic transcription factor 2|Cyclic AMP-responsive element-binding protein 3-like protein 2|BBF2 human homolog on chromosome 7|CR3L2_HUMAN|CREB3L2
- Antigen Name:B ZIB transcription factor
- ELISA:Yes
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



