Anti-NMDAR2B Rabbit Polyclonal Antibody
Catalog # 77054-814
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:AW490526
- Antigen Symbol:NMDAR2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13224
- Antigen Synonyms:Glutamate receptor, ionotropic, NMDA2B (epsilon 2)|Glutamate Receptor Ionotropic N Methyl D Aspartate subunit 2B|Glutamate [NMDA] receptor subunit epsilon-2|Glutamate [NMDA] receptor subunit epsilon 2|Glutamate receptor subunit epsilon 2|Glutamate receptor ionotropic NMDA2B|GRIN 2B|EIEE27|Glutamate Receptor Ionotropic N Methyl D Aspartate 2B
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:165 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NMDAR2B (amino acids 1419-1468)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77054-814
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NMDAR2B for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: NMDAR2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat