Mouse monoclonal (ABT-FSH) antibody to Follicle Stimulating Hormone for IHC with samples derived from Human.
- Validated Applications: IHC
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: FSH beta 2
Clonality: Monoclonal
Clone: ABT-FSH
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77046-424
- 77046-426
- Antigen Symbol:
- FSH beta 2
- FSH beta 2
- Antigen Name:
- Follicle stimulating hormone subunit beta
- Follicle stimulating hormone subunit beta
- Antigen Synonyms:
- FSH B|follitropin|FSH beta 2|follicle stimulating hormone, beta polypeptide|Follitropin beta chain precursor|FSH beta
- FSH B|follitropin|FSH beta 2|follicle stimulating hormone, beta polypeptide|Follitropin beta chain precursor|FSH beta
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-FSH
- ABT-FSH
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P01225
- UniprotID# P01225
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Follicle-Stimulating Hormone
- Synthetic peptide derived from human Follicle-Stimulating Hormone
- Form:
- Liquid
- Liquid
- Purification:
- Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Storage Buffer:
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0.5% Bovine Serum Albumin and 0.1% Kathon CG/ICP
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested Applications:
- IHC
- IHC